1-Methylpyrrolidine ≥95%

Supplier: Ambeed
Danger

Synonyms: N-Methylpyrrolidine

A147130-25G
77549-808EA 14.42 USD
77549-808
1-Methylpyrrolidine ≥95%
1-Methylpyrrolidine

Caution: for Research Use Only

Formula: C₅H₁₁N
MW: 85.15 g/mol
MDL Number: MFCD00003173
CAS Number: 120-94-5
UN: 2924
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR