Benzyltrimethylammonium hydroxide 25% in water

Supplier: Ambeed
Danger

A1460653-25G A1460653-100G
77549-786EA 15 USD
77549-786 77597-296
Benzyltrimethylammonium hydroxide 25% in water
Benzyltrimethylammonium hydroxide

Caution: for Research Use Only

Formula: C₁₀H₁₇NO
MDL Number: MFCD00008281
CAS Number: 100-85-6
UN: 3267
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR