Rhodamine 6G 90 HPLC_ASSAY_METHOD

Supplier: Ambeed
Warning

Synonyms: 9-(2-(Ethoxycarbonyl)phenyl)-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride, 9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride

A1372003-25G A1372003-100G A1372003-500G
77549-666EA 12 USD
77549-666 77621-908 77621-910
Rhodamine 6G 90 HPLC_ASSAY_METHOD
Rhodamine 6G

Caution: for Research Use Only

Formula: C₂₈H₃₁ClN₂O₃
MW: 479.02 g/mol
MDL Number: MFCD00012665
CAS Number: 989-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR