Dodecyltrimethylammonium chloride 97%

Supplier: Ambeed
Danger

Synonyms: (1-Dodecyl)trimethylammonium chloride, N,N,N-Trimethyldodecan-1-aminium chloride, DTAC

A122658-25G A122658-100G A122658-500G
77549-444EA 17 USD
77549-444 77566-320 77599-770
Dodecyltrimethylammonium chloride 97%
Dodecyltrimethylammonium chloride

Caution: For research use only.

Formula: C₁₅H₃₄ClN
MW: 263.89 g/mol
MDL Number: MFCD00041974
CAS Number: 112-00-5
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR