L(+)-Potassium sodium tartrate, Wasser_7 99% hydrate

Supplier: Ambeed
Warning

Synonyms: (R,R)-(+)-Potassium sodium tartrate, L(+)-Tartaric acid potassium sodium salt, Potassium sodium L(+)-tartrate, L-Potassium sodium tartrate, (R,R)-(+)-Tartaric acid potassium sodium salt, (+)-Potassium sodium L-tartrate

A114587-1000G A114587-100G A114587-500G
77549-238EA 62 USD
77549-238 77549-240 77549-242
L(+)-Potassium sodium tartrate, Wasser_7 99% hydrate
L(+)-Potassium sodium tartrate

Caution: for Research Use Only

Formula: C₄H₄KNaO₆
MW: 210.16
Density: 1.79
MDL Number: MFCD00065391
CAS Number: 304-59-6
Merck Index: 12,07840

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR