Cell Culture Plates
Supplier: NEST SCIENTIFIC
712002 712003 712011 713001 713011 714001 714011 748001 748002 748003 748011 701001 701002 701003 701011 701101 701111 701201 701211 701301 701311 702001 702002 761001 761011 761301 761311 761601 761611 712001 702003 702011 703001 703002 703003 703011
77508-588CS 93.36 USD
77508-588 77508-590 77508-592 77508-594 77508-596 77508-598 77508-600 77508-730 77508-732 77508-734 77508-736 77508-504 77508-506 77508-508 77508-510 77508-512 77508-514 77508-516 77508-518 77508-520 77508-522 77508-524 77508-526 77508-754 77508-756 77508-758 77508-760 77508-762 77508-764 77508-586 77508-528 77508-530 77508-532 77508-534 77508-536 77508-538
Cell Culture Plates
Microplates Cell Culture Plates
NEST cell culture plates have round edges for better handling. Low profile, low-evaporation lid design assures uniform growth.
  • Sterilized by E-beam, SAL=10⁻⁶
  • High clarity, 100% virgin polystyrene
  • TC refers to vacuum plasma tissue culture treatment
  • Clear lot number for batch traceability
  • Markings of well coordinates available for 12, 24, 48 and 384 well plates
  • Non-pyrogenic, DNase/Rnase free
Tissue treated and non-treated are both available. Non-treated growth surface is more hydrophobic than tissue culture surface and are similar to the surface of a bacteriological Petri dish. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens.
Grooves are designed at the edge of the culture plate to avoid the edge effect to the greatest extent and to ensure that the cells maintain their optimal state during cell culture.