Orbital Shaker, CO₂ Resistant
Supplier: Nest Scientific
105008
77507-388CS 3321.13 USD
77507-388
Orbital Shaker, CO₂ Resistant
Shakers and Mixers Orbital Shakers
This product is developed for it to be used in high humidity and CO₂ environment. Also, it can be used as a common shaker. Its driven mechanism is composed of magnets and sealed electrical coils. It is maintenance free and has long life time.
  • Orbital shaking through magnetic power eliminates the need to replace belts
  • Small footprint ideal for use in a CO₂ incubator
  • Specially treated mechanical components protect up to 20% CO₂ and 95% humidity conditions
  • Minimal heat dissipation assures conditions within the CO₂ chamber are not effected
  • CO₂ resistant shaker can be used outside of CO₂ incubator for maximum value
  • Humidity: <99% RH
Convenient external control box with LED display allows for easy adjustments without opening the chamber door. The controller can be detached from the body of the shaker so that its speed can be controlled remotely.
It can hold 30×50 ml, 15×100 ml, 15×150 ml, 15×200 ml, 15×250 ml, 9×500 ml, 6×1000 ml, 4×2000 ml, 2×3000 ml, 1×5000 ml standard flasks.