Ultrasonic Cleaning Solutions, Branson
Supplier: Branson Ultrasonics
000-955-114 000-955-014
21830-120CS 325.1 USD
21830-120 21830-200 21830-356 21830-390 21830-083 89065-251 76239-696
Ultrasonic Cleaning Solutions, Branson
Cleaning Agents Detergents and Surface Cleansers
For use with ultrasonic cleaners
  • All cleaning solutions dilute with water
  • Biodegradable
MC-1 metal cleaner is a phosphate and caustic-free alkaline cleaner formulated for heavy duty industrial use. It removes oils, greases and a wide variety of soils from aluminum and aluminum alloys, as well as copper, brass, and steel substrates. Cleaner is also effective in removing stamping and cutting lubricants and light polishing media. It displaces soil and floats it to the surface for easy removal with a skimmer.
General purpose cleaning solution removes general soil, fingerprints, dust, tissue, and light oils. Industrial strength solution removes oil, metal chips, grease, and carbon. Oxide remover solution removes rust and metal oxides.
MC-2 metal cleaner removes oils, greases and a wide variety of soils from ferrous metals, steel alloys, titanium alloys, copper and copper alloys and stainless steel (not recommended for aluminum and aluminum alloys).
Caution: Solutions cannot be used with aluminum products, with the exception of 89065-250.