Courier 7000 Shipping Scales
Supplier: Ohaus
30745875 30745876 30745877 30745878 30745879
77321-494EA 637.43 USD
77321-494 77321-496 77321-498 77321-500 77321-502
Courier 7000 Shipping Scales
Balances and Scales
Accurate standard scales for specific shipping applications.
  • Designed to withstand constant use from offices to distribution centers
  • Fast setup with connection to shipping systems like UPS and FedEx
  • 'GeoCode' application for local calibration adjustment
Display hold retains weight data and presents it on a large, easily accessible backlit display. Its weight alert mode alerts users to packages that may be too heavy in relation to defined values.