Fischer Technical Deluxe Classroom Vacuum Kits
Supplier: Fischer Technical
LAV-DKIT LAV-3/G-DKIT
470316-944EA 363.65 USD
470316-944 470348-974
Fischer Technical Deluxe Classroom Vacuum Kits
Pumps Vacuum Pumps
Demonstrate many exciting low-pressure phenomena in your physical science classroom.
  • Perfect for chemistry and physics applications
  • Safe at high vacuum pressures
This kit comes with everything you need to achieve high vacuum. Demonstrate water boiling at room temperature as well as what happens to a balloon at high vacuum, plus much more!
Delivery information: Kit includes (1) acrylic pump plate, (1) 6×11" glass bell jar, 5' high vacuum tubing, (2) hose clamps, (1) quart high vacuum pump oil, (3) balloons, and (1) petri dish. Your choice with or without LAV-3/G high vacuum pump.