SilTite® μ-Unions, Gas Chromatography Connections
Supplier: Thermo Scientific
290SM404 290SM405 290SM321 290SM322 290SM301 290SM302 290SM303 290SM304 290SM305 290SM401 290SM403
10057-276EA 219.14 USD
10057-276 10057-278 10057-282 10057-284 10057-448 10057-450 10057-452 10057-454 10057-456 10057-460 10057-464
SilTite® μ-Unions, Gas Chromatography Connections
Fittings and Connectors
The SilTite μ-Union is a connector for GC capillary columns, giving zero dead volume. The product has low thermal mass - it is only 9 mm in length and has a mass <0.5 g. It is available in kits to connect columns from 0.1 to 0.53 mm ID.
  • Zero dead volume - giving optimized peak shapes
  • FingerTite technology - easy to install and leak-free
  • Highly inert and robust
Ordering information: Kit contains 5 × ferrules, 2 × union fittings, installation jigs, installation instructions.