HelixMark® Standard Platinum-Cured Silicone Tubing, Momentive Material, Freudenberg Medical
Supplier: Freudenberg Medical
HelixMark®
60-805-25 60-805-01 60-805-05 60-805-26 60-805-31 60-805-33 60-805-34 60-805-29 60-805-06 60-805-04 60-805-03 60-805-10 60-805-06 60-805-25 60-805-04 60-805-22 60-805-07 60-805-24 60-805-02 60-805-28 60-805-02 60-805-32 60-805-29 60-805-24 60-805-10 60-805-30 60-805-33 60-805-03 60-805-05 60-805-22 60-805-27 60-805-34 60-805-26 60-805-21 60-805-08 60-805-31 60-805-09 60-805-08 60-805-11 60-805-28 60-805-23 60-805-09 60-805-32 60-805-30 60-805-01 60-805-27 60-805-23 60-805-11 60-805-07
55566-064EA 478.47 USD
CA55566-074 55566-064 CA55566-072 55566-062 CA55566-070 55566-060 CA55566-078 55566-068 CA55566-076 55566-066 55566-090 55566-098 55566-054 CA55566-062 55566-052 55566-096 CA55566-060 55566-050 55566-094 55566-092 CA55566-068 55566-058 CA55566-066 55566-056 CA55566-064 CA55566-096 CA55566-052 55566-086 CA55566-094 CA55566-050 55566-084 CA55566-092 55566-082 CA55566-090 55566-080 CA55566-058 CA55566-056 CA55566-054 CA55566-098 55566-088 CA55566-084 55566-074 CA55566-082 55566-072 CA55566-080 55566-070 CA55566-088 55566-078 CA55566-086
HelixMark® Standard Platinum-Cured Silicone Tubing, Momentive Material, Freudenberg Medical
Tubing Transfer Tubing
HelixMark® platinum-cured silicone tubing meets the needs of a broad range of applications and is universally applicable for the transfer of fluids under standard requirements.
  • Fully autoclavable and sterilizable
  • Ideal for use in transfer operations as well as other applications requiring non-leachable material
  • Material certification in every package
  • Lot traceability
HelixMark® tubing meets USP Class VI and EP 3.1.9 biocompatiblity requirements. It is manufactured in Freudenberg Medical's FDA registered medical device facility following Good Manufacturing Practices (GMPs) in accordance with The Quality Systems Requlations.
Durometer hardness: Shore A, 50. Tensile strength: 1300 psi.
Certifications: Certified to meet ISO 13485 quality standards. Meets or exceeds USP Class VI biocompatibility requirements.
Delivery information: Tubing is packaged double-bagged in crush-resistant boxes with material certification and lot traceability included in every box.
Packaging: Tubing is individually packaged in 50 foot coil.