Citric acid in ACS Reagent grade water

Supplier: RPI
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

C34525-500.0 C34525-3000.0 C34525-5000.0 C34525-1000.0
76345-416EA 39.4 USD
76345-416 77510-212 77510-214 77510-210
Citric acid in ACS Reagent grade water
Citric acid

Caution: Research or further manufacturing use only, not for food or drug use.

Formula: HOC(COOH)(CH₂COOH)₂
Melting Pt: 153...159 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00011669
CAS Number: 77-92-9

Order Now

Specification Test Results

Appearance White Crystals
IR spectrum Conforms to structure
Purity (Titration by NaOH) ≥99.50%
Chloride Content ≤0.001%
Insoluble Matter ≤0.005%
Iron ≤3 ppm
Lead ≤2 ppm
Oxalate Pass
Phosphate ≤0.001%
Residue on Ignition (Ash) ≤0.02%
Subs Carbonizable by Hot H2SO4 Pass
Sulfur Compounds ≤0.002%
Grade ACS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR