Cystine Heart Agar

Supplier: RPI

C40010-1000.0 C40010-2000.0 C40010-500.0 C40010-5000.0
76345-438EA 304.53 USD
76345-438 76345-440 76345-442 77510-218
Cystine Heart Agar
Media for Microbiology Dehydrated Media for Microbiology

Supports excellent growth of gram-negative cocci and other pathogenic organisms. With added hemoglobin it is used for cultivation of Francisella tularensis. Prepare medium by mixing 51 grams of agar medium in 1 Liter of purified water. Mix, heat and autoclave. Cool to 50 to 60 °C. Add 100 ml sterile 2% hemoglobin solution. Dispense into sterile Petri dish or tube.


  • Appearance: Homogeneous Free Flowing, Beige Powder
  • Microorganism (Neisseria meningitides ATCC 13090): Fair to Excellent
  • Microorganism (Staphylococcus Aureus ATCC 25923): Fair to Excellent
  • Microorganism (Streptococcus pneumoniae ATCC 6305): Fair to Excellent
  • Agar: 15.0 g/L
  • Beef Heart Infusion: 10.0 g/L
  • Dextrose: 10.0 g/L
  • Proteose Peptone: 10.0 g/L
  • Sodium Chloride: 5.0 g/L
  • L-Cystine: 1.0 g/L


Formula per Liter:
Beef Heart Infusion: 10 g
Proteose Peptone: 10 g
Dextrose: 10 g
Sodium Chloride: 5 g
L-Cystine: 1 g
Agar: 15 g
Shipping Weight: 1.2 lbs.


Caution: Research or further manufacturing use only, not for food or drug use.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR