Uvex Strategy® Goggles, Honeywell Safety
Supplier: Honeywell Safety Products
S3810 S3805 S3800 S3815
89013-228CS 1711.11 USD
89013-228 89013-232 89013-226 89013-224 89013-230
Uvex Strategy® Goggles, Honeywell Safety
Goggles
Uvex StrategyUvextreme® goggles offer all-day comfort, exceptional fit, and reliable protection.
Goggles are available with four ventilation options to meet most work applications: direct vent, closed vent, indirect vent, and indirect vent with foam lining to protect against dust, sand, or small particle incursion.
The lightweight, flexible TPE body conforms to the face, ensuring a snug fit and maximum protection in the harshest working conditions. The optically perfected polycarbonate lens delivers excellent peripheral vision and fits comfortably over most prescription eyewear. Lenses offer 99.9% UV protection and feature UvextraUvextreme® AF coating to minimize fogging. Goggle headbands are available in standard fabric or neoprene for chemical applications.
Certifications: Goggles comply with ANSI Z87+ and CSA Z94.3 standards.