RPR Carbon Antigen Suspension, ASI
Supplier: ARLINGTON SCIENTIFIC INC.
901040 901010 987333
80087-334EA 1088.98 USD
80087-334 80087-330 80087-332
RPR Carbon Antigen Suspension, ASI
Clinical Diagnostic Supplies Clinical Reagents and Kits
Carbon Antigen for the use with Arlington Scientifc RPR Syphilis card test
  • Screw cap vials - No glass ampules
  • Made with pride in the USA
  • Longest shelf life - up to 24 months
  • Proven results on CAP, AAB, and API Surveys
Certifications: CPT Code 86592. 510(k)-K851504. CE Marked.