Rubella Virus Antibody Test Kits, ASI

Supplier: ARLINGTON SCIENTIFIC INC.

600500 600100 600030
80087-328EA 3584.4 USD
80087-328 80087-324 80087-326
Rubella Virus Antibody Test Kits, ASI
Clinical Diagnostic Supplies Clinical Reagents and Kits

The ASI Rubella (also known as German Measles) test is a convenient and specific rapid latex particle agglutination test for the Qualitative and Semiquantitative determination of rubella virus antibodies in serum. The kit can be used to test diluted or undiluted samples. The test aids in the diagnosis of recent or active rubella infection and in the determination of immune status.


  • Excellent reproducibility and outstanding accuracy
  • Qualitative and semiquantitative results
  • 3 Levels of liquid, ready-to-use controls included
  • Results in 8 minutes
  • White latex agglutination test
  • Correlation with gold standard - Hemagglutination Inhibition Test (HAI)
  • Proudly made in USA
  • Longest shelf life - up to 18 months
  • Sensitivity: 1-2 IU/mL (undiluted);10-20 IU/mL (diluted)
  • Specificity: >99%


Certifications: CE marked. CPT Code 86762. 510(k)-K031490

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR