RapidQC® Complete Quality Control and Assurance Materials, Siemens Healthineers
Supplier: Siemens Healthineers
RapidQC® Complete Quality Control and Assurance Materials from Siemens Healthineers are for in vitro diagnostic use for the quality control of pH/blood gas analyzers, ISE analyzers, CO-oximeters, and metabolite analyzers.
- Unique formulation
- Three levels of complete solution
This complete solution is a tri-level quality control material, uniquely formulated to provide multiple control parameters and values for pH/blood gas analyzers, ISE analyzers, CO-oximeters, and metabolite analyzers. There are three levels of complete solution, each provided in 2.5 mL ampules. Three levels allow verification of analyzer performance at several points in the clinical range. The complete solution is a buffered bicarbonate solution containing Na+, K+, Ca++, Cl-, carbon dioxide, oxygen, nitrogen, glucose, lactate and dyes.