L-Selenomethionine ≥97% (by HPLC), solid, Calbiochem®, Millipore®

Supplier: MilliporeSigma
Danger

Synonyms: L-SelenoMethionine, (S)-2-Amino-4-(methylseleno)butyric Acid, H-Se-Met-OH

M-Clarity™ quality level = MQ100

561505-1GM 561505-250MG
80601-460EA 806.42 USD
80601-460 80055-444
L-Selenomethionine ≥97% (by HPLC), solid, Calbiochem®, Millipore®
L-Selenomethionine

Increases the expression of glutathione peroxidase.

Formula: C₅H₁₁NO₂Se
Storage Temperature: Ambient
MDL Number: MFCD00037210
CAS Number: 3211-76-5
UN: 3283
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR