Diethyl pyrocarbonate

Supplier: RPI
Warning

D43060-200.0 D43060-25.0 D43060-5.0 D43060-100.0
76345-804EA 846.44 USD
76345-804 76345-806 76345-808 77510-250
Diethyl pyrocarbonate
Diethyl pyrocarbonate

Modification reagent for His and Tyr residues in proteins. Inhibitor of nucleases. DEPC treated water is used in handling of RNA in the laboratory to reduce the risk of RNA being degraded by RNases.


Caution: Research or further manufacturing use only, not for food or drug use.

Formula: O(COOC₂H₅)₂
Storage Temperature: Refrigerator
MDL Number: MFCD00009106
CAS Number: 1609-47-8
EINECS: 216-542-8

Order Now

Specification Test Results

Appearance Clear, Light Yellow Liquid
Density @20°C 1.116 - 1.126 gm/ml
Identification Pass
Purity ≥ 97.5 %
Refractive Index @20°C 1.393 - 1.403


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR