Dextran sulfate sodium salt
Supplier: RPI
|
Warning
|
D20020-10.0
D20020-100.0
D20020-1000.0
D20020-50.0
D20020-250.0
D20020-500.0
76345-746EA
42.26
USD
76345-746
76345-748
76345-750
76345-752
77510-236
77510-238
Dextran sulfate sodium salt
Dextran sulphate sodium salt
Caution: Research or further manufacturing use only, not for food or drug use.
Formula:
(C₆H₇Na₃O₁₄S₃)n |
MDL Number:
MFCD00081551 CAS Number: 9011-18-1 |
Specification Test Results
Appearance | White to off-white crystalline powder |
Identification | Pass Test |
Loss on Drying | ≤ 10 % |
Free Sulfate | ≤ 0.5 % |
pH (1%, Water) @ 25oC | 5.0 - 7.5 |
Chloride | ≤ 1000 ppm |
Sulfur Content | 17.0 - 20.0 % |
Molecular Weight | 500000 |
BSE/TSE Free | BSE/TSE Free |
Country of Origin | Report |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...