Colchicine Colchicum autumnale, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 7α-H-Colchicine, (S)-(-)-Colchicine

234115-1GM 234115-5GM
80055-174EA 112.23 USD
80055-174 80501-026
Colchicine Colchicum autumnale, Sigma-Aldrich®
Colchicine

Inhibitor of mitosis used in cell division studies.


Disrupts microtubules and inhibits tubulin polymerization. Induces apoptosis in PC12 cells and in cerebellar granule cells.

Formula: C₂₂H₂₅NO₆
MW: 399.44 g/mol
MDL Number: MFCD00078484
CAS Number: 64-86-8
Merck Index: 12,02536

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR