2-Propanol ≥99.90%, B&J Brand™ for applications requiring low water content, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Isopropyl alcoholIsopropyl Rubbing Alcoholsec-PropanolIsopropyl rubbing alcohol2-Hydroxy propanesec-Propyl AlcoholDimethyl carbinol2-HydroxypropanePropane, 2-hydroxy-Propan-2-olIPAIsopropanol

324-6X1L
BJ324-6X1LCS 590.74 USD
BJ324-6X1L
2-Propanol ≥99.90%, B&J Brand™ for applications requiring low water content, Burdick & Jackson™
2-Propanol

Isopropyl Alcohol B&J Brand™ for applications requiring low water content


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 77…83 °C (1.013 hPa)
Melting Pt: –90 °C
Density: 0.784…0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II

Order Now

Specification Test Results

gC Analysis 99.9 % min.
refractive Index (20ºC) 1.3766 - 1.3778
residue 1 mg/L max.
uV Absorbance @ 225 nm 0.16 AU max.
uV Absorbance @ 250 nm 0.02 AU max.
uV Absorbance @ 300 nm 0.005 AU max.
uV Absorbance @ 400 nm 0.005 AU max.
uV Cutoff 205 nm max.
water by Karl Fischer Titration 0.01 % max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR