Sight Savers® Premoistened Antifog Lens Cleaning Tissues, Bausch & Lomb®
Supplier: Bausch & Lomb
Sight Savers®
8576
52848-800CS 446.22 USD
52848-800
Sight Savers® Premoistened Antifog Lens Cleaning Tissues, Bausch & Lomb®
Wipes Disposable Wipes and Towels Lens Papers
These premoistened silicone-based antifog and antistatic tissues are ideal for cleaning uncoated lenses, and other glass and plastic applications.
  • Tissues work in both cold-to-hot and hot-to-cold conditions
  • Unique package design allows easy dispensing of large 12.7×0.3 cm (5×8") folded tissues
  • Cardboard dispenser box measures 25.4H×12.4W×5.3D cm (10×4⁷/₈×2¹/₈") and contains 100 tissues