Sulfuric acid 10% (w/w)
Supplier: Ricca Chemical
|
Danger
|
R8153000-1A
RCR8153000-1AEA
54.11
USD
RCR8153000-1A
Sulfuric acid 10% (w/w)
Sulfuric acid
Formula:
H₂SO₄ MW: 98.08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 UN: 2796 ADR: 8,II |
Specification Test Results
Appearance | Colorless liquid |
Assay (vs. Sodium Hydroxide/Phenolphthalein) | 9.95-10.05 % (w/w) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...