Mordant black 11 triturate solution R 1% (w/w) in sodium chloride Reag. Ph. Eur.
Supplier: Ricca Chemical
R5215000-100A6
RCR52150-100A6EA
80.95
USD
RCR52150-100A6
Mordant black 11 triturate solution R 1% (w/w) in sodium chloride Reag. Ph. Eur.
Mordant black 11 triturate
Specification Test Results
Appearance | Gray solid |
Suitability for Use | Wine Red (excess Ca/Mg) - Pure Blue (excess EDTA) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)