Diphenylamine 1% (w/v) in sulfuric acid

Supplier: Ricca Chemical
Danger

Synonyms: N-PhenylbenzeneamineN-Phenylaniline

R2584000-120C
RCR2584000120CEA 67.85 USD
RCR2584000120C
Diphenylamine 1% (w/v) in sulfuric acid
Diphenylamine
Formula: (C₆H₅)₂NH
MW: 169.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003014
CAS Number: 122-39-4
UN: 1830
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR