Diammonium hydrogen citrate solution USP >251< for lead
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Ammonium citrate dibasic
, Diammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate
, Citric acid diammonium salt
R0627920-500A
RCR0627920500AEA
113.27
USD
RCR0627920500A
Diammonium hydrogen citrate solution USP >251< for lead
di-Ammonium hydrogen citrate
Formula:
HOC(CO₂H)(CH₂CO₂NH₄)₂ MW: 226.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013068 CAS Number: 3012-65-5 |
Specification Test Results
Appearance | Colorless to slight yellow |
pH at 25°C (Method: SQCP027, SQCP033) | 8.0-9.0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...