Petri Dishes
Supplier: GSC International
1500-7-10 1500-8-10 1500-9-10 1500-10-10 1500-3-CS 1500-4-CS
470338-642CS 65.3 USD
470338-642 470338-724 470338-702 470338-694 470338-664 470338-626
Petri Dishes
Culture Dishes Petri Dishes
In a science lab or classroom, a petri dish is used to culture different types of cells, including bacteria and molds.
  • Durable and resistant to cracks and scratches
  • Not autoclavable
Get the reliable Petri dish you need for your scientific work. Crafted from high-quality soda lime glass, our Petri dishes are built to last, resistant to cracks and scratches for dependable performance. While they may not be autoclavable, the two-piece design with a lid and bottom dish provides ample space and easy access to protect your cell cultures.
Order this caseMore Product Information