Flint Glass Tubings
Supplier: GSC International
7MMSLT-24 10MMSLT-48
470338-614EA 11.8 USD
470338-614 470346-690
Flint Glass Tubings
Tubing Transfer Tubing
These tubes are perfect for replacement pieces in your science lab.
  • Wall thickness of approximately 1 to 1.2 mm
  • Non-polished cut ends
Looking for reliable glass tubing that won't break the bank? This soda-lime variety has a lower melting point than borosilicate, making it a budget-friendly option for your science, education, or DIY needs.
With a wall thickness of around 1 to 1.2 mm, this glass tubing offers a sturdy construction. And unlike some other options, the ends come unfinished, so you can customize them as needed. Sure, the raw edges may not be as polished, but that just means more flexibility for your projects.
Whether you're stocking up your lab, setting up a STEAM classroom, or tackling a home experiment, our Flint Glass Tubing delivers quality and value. It may not be the fanciest, but it gets the job done without draining your wallet. Give it a try and see the difference honest, functional materials can make.