Precision Balances, XPR
Supplier: Mettler Toledo
30875596 30875678
77507-140EA 12332.95 USD
77507-140 77507-142
Precision Balances, XPR
Balances and Scales
Exceptional results and smart quality assurance. 12.1 kg capacity; 0.1 g readability; integrated methods; results notepad; large weighing pan; user passwords; 4× USB; 1× LAN; error-free data transfer; easy cleaning; LabX compatible.
  • Outstanding weighing performance
  • Audit-proof results every time
  • Management of users and routine tests
  • Consistent workflows to simplify tasks and ensure consistent, efficient workflows
  • Safe and easy cleaning
  • Smart design simplifies weighing
The MonoBloc™ weighing cell delivers fast, accurate and reliable results, ensuring high productivity even in challenging weighing conditions. The StatusLight™, LevelControl, and GWP Approved provide at-a-glance information to verify your results are valid and ready for audit any time. Add your test schedules to the balance to be notified when tests are due. User profiles and passwords provide additional process security.
Interfaces:
USB-B (to device)
RS232 (integrated/optional)
USB-A (to device)
Bluetooth (optional)
Ethernet (LAN)
Compliance options:
Data Integrity
Log history (21 CFR Part 11 Compliant)
Password protection
Log history (Basic Metadata)