Primate Enrichment Treats, Acacia Gum, NF/FCC
Supplier: Bio-Serv
F7015 F7015
76325-044BG 63.83 USD
76325-044 CA76325-044
Primate Enrichment Treats, Acacia Gum, NF/FCC
Animal Supplies Animal Environmental Enrichment
Made from the sap of acacia trees. Acacia gum should be a moist and sticky consistency, but not runny, for easier application.
  • Natural form of edible enrichment
  • Add foraging treats and bits for added interest
  • Place in toys and wood sticks to encourage foraging behavior
Shelf life: 12 months, cool dry storage.
Bio-Serv's flavored PRANGTM can be substituted for the water to provide added variety.