Dual-Compartment Dispenser, AK®
Supplier: AK
AK-1478
32799-016EA 563.21 USD
32799-016
Dual-Compartment Dispenser, AK®
PPE Dispensers
Designed to keep an assortment of disposable products in one location, this large-capacity dispenser features two compartments
All-purpose dispenser is made of clear 6 mm (¹/₄") acrylic and is ideal for shoe covers, gloves, bouffants, hairnets, beard covers, and hoods. Openings feature radius edges for a smooth finish.
Compartment dimensions: 24.1× 30.5×45.7 cm (9.5×2×18")