Somso® Glomerulus
Supplier: MARCUS SOMMER
Somso®
LS 7
470162-890EA 318.59 USD
470162-890
Somso® Glomerulus
Models Human Biology Models
Somso® premium quality models.
  • 5 year warranty
Enlarged approx. 700 times, in SOMSO-Plast®. The model shows the arteriola afferens and the arteriola efferens, the capillaries of the glomerulus, urinary pole and Bowman's capsule. In one piece. Mounted on a green board.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.