Kahnetics® Shot Meters, Cooper Tools
Supplier: Cooper Tools
KDS834A KDS824A KDS808
37695-496EA 2764.01 USD
37695-496 37698-828 37697-816
Kahnetics® Shot Meters, Cooper Tools
Manometers
Shot meters are designed for the benchtop and offer easy reading of dials and convenient adjustment of air pressure and vacuum
Economy and Deluxe shot meters also feature digital timer control for metered shot repeatability, shot time durations that are adjustable from 0.01 to 99.99 seconds, and an adjustable four-position push button timer switch. Deluxe shot meters have two 0–100psi regulators and gauges with locking ring; all other shot meters have one 0–100psi regulator and gauge with locking ring. Deluxe shot meters feature an accessory outlet for connection to an external tank.
Each shot meter has a vacuum feature to prevent dripping, an electrically actuated foot-pedal, and a slide switch that easily changes the unit from 115V to 230V. All shot meters are low voltage for safety.