Kahnetics® Foot Valves, Cooper Tools
Supplier: Cooper Tools
Kahnetics®
KDS806 KDS806V
37698-896EA 1118.03 USD
37698-896 37698-900
Kahnetics® Foot Valves, Cooper Tools
Fluid Adhesive Dispensers and Pistons
Low maintenance foot valves have basic on/off function, 0–100psi regulator, and gauge
“Eyeball” accuracy. Foot valves work on compressed air input only; no electricity is required. Foot valve 37698-900 includes a drip-free vacuum accessory.