Liebig Condenser, Ace Glass
Supplier: Ace Glass
9258-02
89074-320EA 185.74 USD
89074-320
Liebig Condenser, Ace Glass
Condensers
Borosilicate glass condenser features an additional vacuum takeoff between top outer joint and jacket
[ST] 14/20 top center and bottom inner joints. Hose connections accept 7.9mm (5/16") I.D. tubing.