Mag-Bind® FFPE RNA 96 Kit
Supplier: OMEGA BIO-TEK
M2551-00
M2551-01
89167-354EA
637.85
USD
89167-354
89167-356
Mag-Bind® FFPE RNA 96 Kit
Nucleic Acid Reagents
Nucleic Acid Purification Kits and Reagents
The Mag-Bind FFPE RNA 96 kit is specially designed for isolation of total RNA from formalin-fixed, paraffin-embedded tissue sections.
The sample is first processed in special buffer by heating to deactivated RNases/DNase and deparaffinization. Then the lysate is treated with Proteinase K to digest proteins. The cleared lysate is then mixed with Mag-Bind particles to capture the capture RNA. After few quick wash steps, purified RNA can be eluted with water or low salt buffer.