Magleads
Supplier: AMERICAN SCIENTIFIC LLC SE
ML300B ML300C ML300R
470355-432PK 20 USD
470355-432 470355-402 470355-440
Magleads
Wire
Simply the easiest and best jumper wires you can have in your classroom.
  • The 4 mm diameter magnets work in all the places that standard clips work
  • No need for an adapter to connect wires to your battery
  • No more fighting with Fahnestock or alligator clips - just click them together!
Instantly make working electrical circuits with these ingenious leads, each fitted with a tiny powerful magnet at either end. Place a couple near to a battery case and they connect by themselves. Then, place them next to a bulb holder connector and you have your first working circuit!