2,5-Dichloroisonicotinic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2,5-Dichloropyridine-4-carboxylic acid
15945-25G
77498-794EA
62.5
USD
77498-794
2,5-Dichloroisonicotinic acid ≥97%
2,5-Dichloroisonicotinic acid
Caution: for Research Use Only
Formula:
C₆H₃Cl₂NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00466640 CAS Number: 88912-26-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...