Supertek® Dual Scale Spring Scales
Supplier: Shiv Dial Sud & Sons
PH10132/1 PH10132/2 PH10132/3 PH10132/4 PH10132/5 PH10132/6 PH10132/7
470353-978CS 951.9 USD
470353-978 470353-982 470353-998 470353-986 470353-980 470353-990 470353-996
Supertek® Dual Scale Spring Scales
Balances and Scales
Plastic spring scales, built to last in any laboratory.
  • Scale calibrated in both grams and Newtons
  • Built-in zero adjustment
  • Crystal clear plastic body for easy viewing
These reliable spring scales are housed in a robust plastic transparent body for easy viewing. Overall length of approximately 285 mm, with upper and lower suspension hooks and zero adjustment. Scales are calibrated in both grams and Newtons.