Phlebotomy Collectors, BD Medical
Supplier: Avantor
305487
BD305635CS 421.57 USD
BD305635 BD305487
Phlebotomy Collectors, BD Medical
Bins Sharps Containers
Preassembled, robust polypropylene sharps collectors with convenient, vertical “point first” drop
For accessories, see BD305451 series.
They feature a patented one-way funnel valve designed to minimize needlesticks and overfilling. Red containers are equipped with natural top with dual access