Jet-melt™ Hot Melt Adhesives, 3M™
Supplier: 3M
7000000879 7100009192
12778-018EA 107.38 USD
12778-018 12778-022
Jet-melt™ Hot Melt Adhesives, 3M™
Adhesives
Solvent-free adhesives achieve bond strength in seconds to bond wood, plastics, foam, fabric, cardboard, and many other surfaces. For use with Polygun™ adhesive applicators (see 12778-012 series).
  • Solvent-free
  • Works in seconds
  • Bonds to many surfaces
Adhesive 12778-018 offers good thermal and electrical properties
Adhesive 12778-022 offers good electrical properties with high strength and heat resistance for potting, wire staking, UL 94 V-O. Amber adhesive has good fuel and oil resistance. Heat resistant to 150°C (302°F).
Noncorrosive to copper.More Product Information