Diamond® Essentials™ Griffin Beakers
Supplier: Globe Scientific
3650-100 3650-10M 3650-1M 3650-250 3650-2M 3650-50 3650-5M 3650-600 3652-100 3652-10M 3652-1M 3652-250 3652-2M 3652-50 3652-5M 3652-600
77447-878PK 28.59 USD
77447-878 77447-880 77447-882 77447-884 77447-886 77447-888 77447-890 77447-892 77447-894 77447-896 77447-898 77447-900 77447-902 77447-904 77447-906 77447-908
Diamond® Essentials™ Griffin Beakers
Beakers
Diamond® Essentials™ griffin style beakers meet the demands for mixing, pouring, stirring, and measuring solutions in many laboratory operations.
  • Manufactured from virgin, USP class VI resins
  • Bold, printed, dual scale graduations
  • Single non-drip spout
  • Autoclavable at 121 °C
  • Chemically resistant to most acids, bases, and many common solvents
These beakers are the safe, shatterproof alternative to glass in operations where plastic is suitable. Value, quality, and performance are insured by precision molding in an automated manufacturing process. The beakers are designed with a single, no drip spout and the beakers with handles facilitate easy pouring.
Available in economic polypropylene (PP) or crystal clear polymethylpentene (PMP).
Certifications: Complies with FDA 21-CFR and European Pharmacopeia.