Sodium nitrate, powder

Supplier: Avantor

ANAC030-025-01
80700-170EA 301.85 USD
80700-170
Sodium nitrate, powder
Sodium nitrate
Formula: NaNO₃
MW: 84.99 g/mol
Melting Pt: 306 °C
MDL Number: MFCD00011119
CAS Number: 7631-99-4

Order Now

Specification Test Results

Assay ≥99.0%
pH 5.5 - 8

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR