ω,ω-Difluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

52569-5G 52569-25G
77494-872EA 133.9 USD
77494-872 77504-372
ω,ω-Difluoroacetophenone ≥97%
ω,ω-Difluoroacetophenone

Caution: For research use only

Formula: C₈H₆F₂O
Storage Temperature: Ambient
MDL Number: MFCD06246879
CAS Number: 395-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR