Nalgene® Boston Round Bottle, Low-Density Polyethylene, Narrow Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
312003-0001 312003-0002 312003-0004 312003-0008 312003-0016 312003-0032
16059-780CS 1287.36 USD
16059-780 16059-782 16059-784 16059-786 16059-788 73521-362
Nalgene® Boston Round Bottle, Low-Density Polyethylene, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
These low-density polyethylene bottles are ideal for packaging, shipping, and storing a variety of laboratory samples
Translucent design allows easy viewing of contents. Highly reliable and durable for long-term use.
Ordering information: Supplied with polypropylene screw caps. Bulk packed.