Leak-Resistant Bottles, Polypropylene, Narrow Mouth, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
209163 209164 209165 209166 209167 209168 209169 209161 209170
16155-163CS 151.08 USD
16155-163 16155-164 16155-165 16155-166 16155-167 16155-168 16155-169 16155-161 66012-882
Leak-Resistant Bottles, Polypropylene, Narrow Mouth, WHEATON®, DWK Life Sciences
Bottles Plastic Bottles
For general laboratory use. Polypropylene bottles feature a special pour lip to facilitate safe, clean dispensing.
  • Double-seal closure for leak-resistance
  • Autoclavable
Note: Before autoclaving, set cap on top of bottle without engaging threads.
Ordering information: Supplied in convenience-packed cartons with polypropylene caps attached.