Productainer BioProcess Containers, Thermo Scientific
Supplier: Thermo Scientific
SH30651.07 SH30651.06
82013-718EA 505.76 USD
82013-718 82013-716
Productainer BioProcess Containers, Thermo Scientific
Bags Media Bags
BioProcess containers are specifically engineered for use in biotechnology and pharmaceutical research and production processes.
  • Containers are top-dispensing
  • Systems are constructed of a medical-grade film that is animal-derived and component-free
Their three-dimensional design achieves optimal filling and conforms better to support containers than traditional pillow-style bags. Each disposable container minimizes the risk of cross-contamination and reduces cleanup and turnaround time in the laboratory. These containers are outfitted with two to five ports and are available in a top- or bottom-dispensing design.
The result is a container highly compatible with a wide range of buffers and process liquids that exhibits outstanding tensile properties, low gas permeability, and low profile of leachable compounds.
Applications include waste collection, bioreactor and fermentation feed and harvest, transport and storage of bulk intermediate product, and formulation and filling of sterile media, buffers, and other process liquids.
Ordering information: All container ports are equipped with tube extensions of varying length and LL (Luer-lock) or QC (quick connect) tubing connectors. 3-D BioProcess Container systems are designed for use with standard outer containers; contact your Avantor representative for more information.