Terrycloth Baker's Pads, Wells Lamont
Supplier: Wells Lamont
B-PAD
80091-619EA 6.43 USD
80091-619
Terrycloth Baker's Pads, Wells Lamont
Hot Hand Protectors
Heat-resistant terry cloth pads are designed for use in food service applications.
  • Two layers of heat-resistant terry cloth
  • Heavyweight material
  • Pads protect against up to 238 °C (460 °F)
  • For use in food service applications
The 24.1 cm long pad is constructed of two layers of terry cloth with a handhole.
All pads feature an elastic loop that allows them to be worn around the wrist or hung.